DS-33 Matrix Standard Kit Dx (Dye Set G5)
Applied Biosystems™

DS-33 Matrix Standard Kit Dx (Dye Set G5)

The DS-33 Matrix Standard Kit Dx (Dye Set G5) is used to generate the multicomponent matrix required when analyzing 6-FAM, VIC, NED, PET, and LIZ dye-labeled DNA fragments on Applied Biosystems Dx genetic analyzers.
Have Questions?
Catalog NumberQuantity
A257758 Runs
Catalog number A25775
Price (USD)
316.00
Each
Quantity:
8 Runs
Price (USD)
316.00
Each

The DS-33 Matrix Standard Kit Dx (Dye Set G5) is used to generate the multicomponent matrix required when analyzing 6-FAM, VIC, NED, PET, and LIZ dye-labeled DNA fragments on Applied Biosystems Dx genetic analyzers. The Data Collection Software for these instruments uses the multicomponent matrix to automatically analyze the five different-colored fluorescent -labeled samples in a single capillary.

For In Vitro Diagnostic Use Only
Specifications
For Use With (Equipment)3500 Dx Genetic Analyzer
Label or DyeLIZ
Product TypeDS-33 Matrix Standard Kit Dx
Quantity8 Runs
Shipping ConditionWet Ice
Software TypeData Collection Software
Starting MaterialDNA
Unit SizeEach
Contents & Storage
Contains 5-dye labeled oligonucleotides used for generating the multicomponent matrix in a 1X TE solution.

Store in refrigerator (2–8°C). Do not freeze.