What cells can be cultured in Exosome-Depleted FBS, and at what concentration?
We have specifically tested Jurkat, MCF-7, HeLa, PC3, HEK293, and A549 cells, all of which exhibited acceptable performance. HeLa cells showed the most sensitivity to the presence of Exosome-depleted FBS. Adjustments may be needed for the concentration of Exosome-depleted FBS to match the requirements of the specific cell line and system. We recommend starting at 10% Exosome-depleted FBS, and adjusting up or down from there as necessary. Also, the Exosome-depleted FBS has been tested and validated for a single passage of cells for 48 hours (and we have also tested HeLa, MCF-7, and HEK293 cells successfully up to 96 hours of culture).