Mini Gel Tank Lid - FAQs
Mini Gel Tank Lid - FAQs
View additional product information for Mini Gel Tank Lid - FAQs (A25944)
5 product FAQs found
Can I prepare my protein sample with the reducing agent and store it for future use?
DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.
Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.
My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?
Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.
Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.
Can I purchase replacement parts for the Mini Gel Tank?
Here are the replacement parts we offer for the Mini Gel Tank:
Replacement part - Cat. No.
Gel Runner Tank - B4478641
Mini Gel Tank Lid - A25944
Mini Gel Tank Base - A25950
Cassette Clamp, left - A25946
Cassette Clamp, right - A25945
Cassette Clamp Cam Handle Set - A26732
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What are the components of the Mini Gel Tank?
The Mini Gel Tank includes the Mini Gel Electrophoresis Tank, Mini Gel Tank Lid, Cassette Clamp (Right), 10 cm, Cassette Clamp (Left), 10 cm, Mini Gel Tank Base, Invitrogen Power Supply Adapters, and Gel Knife.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.