Product Image
Mini Gel Tank Base
The Mini Gel Tank Base is a replacement part for the Mini Gel Tank. It holds the Gel Runner Tank.AboutRead more
Have Questions?
Catalog NumberQuantity
A259501 unit
Catalog number A25950
Price (USD)
44.65
Online exclusive
45.82
Save 1.17
Each
Quantity:
1 unit
Price (USD)
44.65
Online exclusive
45.82
Save 1.17
Each
The Mini Gel Tank Base is a replacement part for the Mini Gel Tank. It holds the Gel Runner Tank.
About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex™ gels, NuPAGE™ gels, and Bolt™ Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt™ Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 unit
Shipping ConditionRoom Temperature
For Use With (Equipment)Mini Gels
Gel CompatibilityNovex™ Mini Gels
Gel SizeMini
TypeMini Gel Tank Base
Unit SizeEach
Frequently asked questions (FAQs)
Can I prepare my protein sample with the reducing agent and store it for future use?
DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.
Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.
My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?
Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.
Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.
What are the storage conditions for Bolt gels?
Similar to NuPAGE gels with storage temperatures of 4 to 25 degrees C.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I purchase replacement parts for the Mini Gel Tank?
Here are the replacement parts we offer for the Mini Gel Tank:
Replacement part - Cat. No.
Gel Runner Tank - B4478641
Mini Gel Tank Lid - A25944
Mini Gel Tank Base - A25950
Cassette Clamp, left - A25946
Cassette Clamp, right - A25945
Cassette Clamp Cam Handle Set - A26732
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What are the components of the Mini Gel Tank?
The Mini Gel Tank includes the Mini Gel Electrophoresis Tank, Mini Gel Tank Lid, Cassette Clamp (Right), 10 cm, Cassette Clamp (Left), 10 cm, Mini Gel Tank Base, Invitrogen Power Supply Adapters, and Gel Knife.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.