CTS™ Immune Cell Serum Replacement (SR), 500 mL - FAQs
CTS™ Immune Cell Serum Replacement (SR), 500 mL - FAQs
View additional product information for CTS™ Immune Cell Serum Replacement (SR) - FAQs (A4702903, A2596101, A4702901, A2596102, A4702902)
2 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
What are the storage conditions for CTS Immune Cell Serum Replacament (SR)? Upon thawing, can I aliquot and refreeze at -20 degrees C?
After completely thawing the initial product, it can be aliquoted and refrozen for another usage. The thawed aliquots are stable for 4 weeks when stored at 2-8 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.