RecoverAll™ Multi-Sample RNA/DNA Isolation Workflow
Invitrogen™

RecoverAll™ Multi-Sample RNA/DNA Isolation Workflow

The RecoverAll™ Multi-Sample RNA/DNA Workflow is a complete workflow package for sequential extraction of RNA and DNA from the sameRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A26069120 Preps
A26135200 Preps
Catalog number A26069
Price (USD)
930.65
Online exclusive
996.00
Save 65.35
Each
Quantity:
120 Preps
Price (USD)
930.65
Online exclusive
996.00
Save 65.35
Each

The RecoverAll™ Multi-Sample RNA/DNA Workflow is a complete workflow package for sequential extraction of RNA and DNA from the same formaldehyde- or paraformaldehyde-fixed paraffin-embedded (FFPE) tissue sample. The recovered RNA and DNA are ready for use in downstream applications such as quantitative real-time RT-PCR and targeted sequencing using the Oncomine™ Cancer Research Panel. This workflow package includes three RecoverAll™ Total Nucleic Acid Isolation kits, elution tubes, and nuclease-free water.

Features include:
• Recovers RNA and DNA in separate eluates from the same FFPE section
• Relies on proven Ambion™ RecoverAll technology
• Processes four 20-μm sections per reaction
• Provides enough reagents to perform 60 RNA and 60 DNA isolations (120 total)

Special workflow provides more information from less sample input
FFPE samples are precious, which often requires the choosing of one extraction method over another to save sample. With the RecoverAll Multi-Sample RNA/DNA Workflow, recovering RNA and DNA from the same FFPE tissue sample in a single workflow eliminates the need to choose which nucleic acid to recover from a particular FFPE section.

First, FFPE slide-mounted or tissue blocks sections are deparaffinized and digested with protease. The samples are then loaded on a filter cartridge; DNA remains bound to the filter cartridge while RNA flows through. To recover RNA, the flow-through is applied to a new filter cartridge, treated with DNase, then eluted from the filter. To recover DNA, the DNA bound to the filter cartridge is treated with RNase A, then eluted from the filter.

Related product
Ambion RecoverAll Total Nucleic Acid Isolation Kit

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)PCR, Northern Blotting, RT-PCR (Endpoint), microRNA Analysis, Real-Time PCR, Sequencing
High-throughput CompatibilityNot High-throughput Compatible
No. of Reactions120 Preps
Product LineAmbion, RecoverAll
Quantity120 Preps
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
Sample TypeFFPE and Fixed Samples, Fixed Samples
TargetTotal RNA, miRNA, Total Nucleic Acids, mRNA, Genomic DNA
Unit SizeEach
Contents & Storage
• 3 RecoverAll Total Nucleic Acid Isolation kits (shipped in two parts: one at room temperature and one at -20°C; store as indicated on box)
• 100 elution tubes
• 100 mL nuclease-free water