Product Image
Thermo Scientific™
3500/3500xL Data Collection Software, v3.1 Upgrade
This product is a software upgrade package. It includes a 3500/3500xL Data Collection Software upgrade CD for upgrade from v2Read more
Have Questions?
Catalog NumberQuantity
A262871 license
Catalog number A26287
Price (MXN)
-
Quantity:
1 license
This product is a software upgrade package. It includes a 3500/3500xL Data Collection Software upgrade CD for upgrade from v2 or V3 to v3.1, 1 licensing kit with single license code, and 1 user document CD.
Data Collection Software v3.1 provides the features and functionality that researchers have been requesting to take their research even further. Powered by the Windows™ 7 operating system, our newest and most powerful data collection software provides:
• The removal of on-instrument hard stops of consumables, yielding increased flexibility and control
• New run modules for Sequencing and Fragment Analysis with the 36-cm array for faster run times and increased flexibility
• An improved user interface with added functionality such as customizable filters and user objects
• Performance check flexibility with your polymer of choice to help improve cost savings and efficiencies
• Updated user interface to recognize new polymer size offering (96-sample pouch) and shelf-life extension of the on-deck consumables (polymers, ABC and CBC buffers)
• A usage statistic report for greater visibility into consumable management and expenses
Additional features include:
• Accuracy—a comprehensive dashboard delivers regular updates of instrument status and remaining days of onboard consumables
• Confidence—improved spatial calibration in quality checks with added tests for peaks exceeding thresholds, helping to provide assurance in instrument performance
• Security—Built-in SAE feature set assists in meeting more stringent requirements
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)3500 Genetic Analyzer
FormatPouch
Operating SystemMicrosoft™ Windows™ 7 SP1 32-bit (recommended)
Product Type3500/3500xL Data Collection Software V3.1 Upgrade
Quantity1 license
Shipping ConditionRoom Temperature
Throughput96 Sample Pouch
Software TypeData Collection Software
Unit SizeEach
Contents & Storage
• 3500/3500xL Data Collection Software v2/v3.0 to v3.1 upgrade CD
• 1 licensing kit with single license code, combined RUO and SAE (Security, Audit, and E-signature)
• 1 user document CD
Frequently asked questions (FAQs)
How can I set up auto-analysis for data in GeneMapper with the 3500 Data Collection Software version 3.x?
Beginning with 3500 Data Collection Software version 3, the 3500 Data Collection Software does not support automatic secondary analysis. This is also indicated in the software release notes. To perform secondary data analysis, you will need to manually import the .fsa file into the GeneMapper software. For optimal performance, we recommend that you install the GeneMapper Software on a stand-alone computer that is not part of the 3500 Genetic Analyzer system.
Find additional tips, troubleshooting help, and resources within our Fragment Analysis Software Support Center.
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?
If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Do I need to upgrade or update any secondary analysis software so that it is compatible with 3500/3500xL Data Collection Software v3.1?
If you are upgrading from 3500/3500xL Data Collection Software v1, new Windows 7-compatible secondary analysis software will be required. For all users, if you are going to be using the 36 cm array for sequencing or the 50 cm array with POP-6 polymer and BigDye Terminator v3.1, you will need to apply patches for Sequencing Analysis Software v6, Variant Reporter Software v2 and SeqScape Software v3. You can download the patches from: http://www.thermofisher.com/datacollection3_1.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Since the 36 cm array is now enabled to do sequencing and fragment analysis, will the 36 cm array be included in the 3500/3500xL system package during purchase?
No, the 36 cm array will not be included in the system package. The standard offering will still be the 50 cm array. If you would like the 36 cm array, it is available for purchase separately.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.