QuantStudio™ 3D Digital PCR Reagent Kit

Catalog number: A26360

Applied Biosystems™  Related applications: Digital PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The QuantStudio™ 3D Digital PCR Reagent Kit is designed to confirm the performance of the QuantStudio™ 3D Digital PCR System either at the time of system installation or as part of future, user-defined system verification procedure using established Applied Biosystems™ consumables, reagents, control DNA, and reference assays. The kit may also be used for training users on the QuantStudio™ 3D Digital PCR System workflow.

The QuantStudio 3D Digital PCR Reagent Kit contains CEPH control DNA, GLA and RNase P reference assays, QuantStudio™ 3D Digital PCR Master Mix v2, and nuclease free water. The kit requires use of QuantStudio 3D™ Digital PCR Chip Kit v2 and is fully-compatible with all QuantStudio 3D™ Digital PCR Instrument hardware and software.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): QuantStudio™
Product Line: QuantStudio™
Product Size: 1 kit
Shipping Condition: Dry Ice

Contents & storage

• 1 mL CEPH Control DNA, 50 μg/mL, store at -20°C.
• GLA Reference Assay, store at -20°C.
• RNase P Reference Assay, store at -20°C.
• Nuclease-free water, store at -20°C.
• 1.5 mL QuantStudio 3D Digital PCR Master Mix v2, store at -20°C until first use and then store at 2–8°C.

Documents

Manuals & protocols

Recommended products