A2644401 - FAQs
A2644401 - FAQs
View additional product information for A2644401 - FAQs (A2644401)
3 product FAQs found
Can RevitaCell Supplement be used in addition to a traditional ROCK Inhibitor?
No. Combination of RevitaCell Supplement with a traditional ROCK inhibitor will result in deleterious effects on PSCs.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
The RevitaCell Supplement in the PSC Cryopreservation Kit has a strong smell. Is this normal?
Yes, it is normal that the RevitaCell Supplement has a strong smell.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between the PSC Cryopreservation Kit and the CTS PSC Cyropreservation Kit?
The CTS PSC Cryopreservation Kit (Cat. No. A4239301) provides the same efficient recovery, consistency, and reduced variability as the PSC Cryopreservation Kit (Cat. No. A2644601). However, these two kits have different intended use statements, i.e., “For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals” and “For Research Use Only”, respectively. Furthermore, the CTS PSC Cryopreservation Kit has full USP sterility testing info and a Drug Master file (DMF), whereas the sterility testing of the PSC Cryopreservation is via a modified USP method, and it does not have a DMF.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.