POROS™ GoPure™ MabCapture™ A Select Pre-packed Column, 1.2 x 10 cm, 11.3 mL

Catalog number:  A26465

Applied Biosystems™  Related applications: Chromatography

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

POROS™ GoPure™ columns combine the flexibility of a traditional packed column with the convenience and reproducibility of a pre-packed ready-to-use column solution. GoPure Pre-Packed Columns speed facility turnaround time by eliminating column qualification testing and the need for equipment cleaning and change-out. In addition, the columns are comparable in cost to packing a traditional column. GoPure Pre-Packed Columns are packed and qualified in a dedicated clean room and are supported by numerous validation studies. Multi-product facilities requiring flexible, efficient, and cost-effective operations will find that GoPure Pre-Packed Columns fill a gap in downstream purification unit operations.

The resin in this GoPure column is POROS™ MabCapture™ A Select Media, providing a high binding capacity matrix for antibody purification.

Convenient—ready-to-run columns speed process setup and facility turnaround time
Flexible design—bed heights available across multiple diameters, allowing columns to be tailored to process requirements
Simplified scalability—qualified, reproducible pre-packed column simplifies process scale up or process modeling
Qualified for use—columns packed in a dedicated clean room and include a column test certificate

Convenience and performance delivered
GoPure Pre-Packed Columns are available in 1.2, 8.0, and 20 cm diameters, each with multiple bed heights that equate to a column volume range of 5 mL to 6.3 L. The simple design is reliable and sanitary. Moreover, the column construction materials meet regulatory requirements for toxicity and being animal origin-free. The columns arrive pre-qualified, sanitized, and stored in a bacteriostatic solution. They are shipped with a column test certificate that includes column efficiency, asymmetry, and permeability measurements.

GoPure Pre-Packed Columns significantly reduce the time required to prepare for chromatography unit operations. In addition, the columns can be campaign-dedicated, eliminating the time usually required to perform and document a cleaning validation. Depending on the facility workload, this time-saving feature can help to expand capacity without capital investments.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Strength: 5% breakthrough capacity > 37 mg/mL Human IgG
Column Type: Affinity
Format: Chromatography Column
Immobilized Phase: Protein A
Product Line: GoPure™, Mabcapture A SELECT™, POROS®
Product Size: 1 column
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products