How can I create a new variant call group in the HRM app on Thermo Fisher Connect, select a color for the variant, and assign wells to that group?
To create a new variant call group in the HRM app on Thermo Fisher Connect, follow these steps:
Find additional tips, troubleshooting help, and resources within our High Resolution Melt Support Center.
- Select the 'Quality Control & Results' tab.
- In the 'Quality Control & Results' pane, select your experiment of interest.
- Select the 'Derivative melt plot'.
- Click 'Actions', and then select 'Manual Call'.
- From the 'Manual Call' dialog box, you have two options:
- To assign the sample to an existing variant call: click 'Select Existing', choose the appropriate call from the 'Group' drop-down menu, and then click 'OK'.
- To create a new variant call: click 'Create New', enter a name for the new call in the 'Group' field, select a color, and then click 'OK'.
- Repeat the above steps to assign more manual calls.
- Click 'Analyze' to reanalyze the data using the manual calls.
Find additional tips, troubleshooting help, and resources within our High Resolution Melt Support Center.