Ion 530™ Chip Kit-4 Reactions
Ion Torrent™
Ion 530™ Chip Kit-4 Reactions
The Ion 530™ Chip Kit-4 Reactions contains 4 barcoded chips for sample tracking and sequencing of 400-600 bp fragments.
Have Questions?
Catalog NumberQuantity
A277634 Chips
Catalog number A27763
Price (MXN)
62,390.75
Each
Quantity:
4 Chips
Price (MXN)
62,390.75
Each
The Ion 530™ Chip Kit-4 Reactions contains 4 barcoded chips for sample tracking and sequencing of 400-600 bp fragments. The kit is intended for use with the Ion 520™ & Ion 530™ ExT Kit-Chef and the Ion S5™ and Ion S5™ XL Sequencing Systems. The Ion 530 Chip electronically detects polymerase-driven base incorporation without the use of fluorescence. By eliminating the use of an optical detection system, this advance in next-generation sequencing technology allows for rapid sequencing times of as little as 4.5 hours for 600 bp sequencing.
The Ion 530 Chip Kit-4 Reactions offers:
  • Sample tracking functionality from an integrated barcode
  • Read lengths of 400-600 bp to support a variety of research applications, including microbial sequencing and metagenomics
  • A rapid sequencing run time of as little as 4.5 hours, generating up to 12 million reads with ExT chemistry
  • Automated handling and loading of templated products for sequencing
  • Compatibility with current library preparation methods
Notes:
This kit is only compatible with the Ion S5 and Ion S5 XL Systems. Use this kit with the Ion 520 & Ion 530 ExT Kit-Chef and Torrent Suite™ Software 5.2 or higher.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Ion GeneStudio S5 System, Ion S5™ XL System
Number of Reads per Chip15-20M
Product LineIon Torrent
Product TypeChip Kit
Quantity4 Chips
FormatKit
Unit SizeEach
Frequently asked questions (FAQs)
How many Oncomine BRCA Research Assay samples can I multiplex on the Ion 530 Chip?
The number of samples that can be multiplexed on the Ion 530 Chip will vary depending on whether automated or manual library preparation is used. If using automated library preparation, 32 somatic samples or 32 germline samples can be multiplexed on the Ion 530 Chip. If using manual library preparation, up to 96 germline samples can be multiplexed on the Ion 530 Chip. This is because of the available Ion Xpress Barcodes.
What is the throughput of the Ion 520 Chip?
For 200 bp sequencing, it will yield 0.6-1.0 Gb. For 400 bp sequencing, it will yield 1.2-2 Gb.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.