Why are the recommended working concentrations different for CTS Vitronectin (VTN-N) Recombinant Human Protein, Truncated vs. Vitronectin (VTN-N) Recombinant Human Protein, Truncated? Are they not the same protein?
These two proteins are the same but have different protein concentrations due to the different methods used to measure the protein concentrations.