Analysis Module Variant Analysis (VA) Software - FAQs
Analysis Module Variant Analysis (VA) Software - FAQs
View additional product information for Analysis Module Variant Analysis (VA) Software - FAQs (A28220)
3 product FAQs found
With the availability of Thermo Fisher Cloud-based software, will you be discontinuing my desktop-based software?
No, all existing desktop-based software will still be available.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the new Sanger modules on Thermo Fisher Cloud?
Quality Check (QC): Allows you to monitor run quality by comparing performance against thresholds
Variant Analysis (VA): Allows you to align reads against a known reference, identify variants against a reference, and report genomic locations and annotations; can be used with multiple amplicons
NGS-to-CE (NGC): Allows you to confirm and validate NGS variants and visualize Venn diagrams between NGS and CE variants
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the benefits of using Thermo Fisher Cloud-based software over desktop-based software?
Using Thermo Fisher Cloud-based software has the following advantages over desktop-based software:
- Can access data using a web browser on a Mac or Windows PC
- Can access data from any location with internet access
- Don’t need a dedicated computer with specific software requirements in order to use it
- No software installation needed
- Scalable computing
- Ability to share data files with multiple users globally
- Secure storage
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.