Attune™ NxT Flow Cytometer, blue/red/violet/green
Invitrogen™

Attune™ NxT Flow Cytometer, blue/red/violet/green

The 2019 Attune NxT Flow Cytometer has expanded functionalities to drive your research forward. Designed and developed to remove commonRead more
Have Questions?
Catalog NumberQuantity
A29001CFR1 each
Catalog number A29001CFR
Price (USD)
-
Quantity:
1 each
The 2019 Attune NxT Flow Cytometer has expanded functionalities to drive your research forward. Designed and developed to remove common barriers associated with flow cytometry, the evolutionary capabilities of the Attune NxT flow cytometer offer adaptable optical configuration options so you can get the most out of your multicolor analysis.

• Free Attune NxT software with researcher-inspired upgrades
• New green laser option for expanded utility and fluorochrome options
• Simple push-button sample recovery function

Freakishly fast flow
The innovative combination of acoustic focusing technology coupled with traditional hydrodynamic focusing results in acquisition speeds up to 10X faster than other flow cytometers. This technology reduces the time needed to obtain results.

• True 35,000 event/sec (+10% coincidence rates)
• Maximum electronic speed of 65,000 events/sec
• Analyze small or limited samples faster than ever
• Efficiently run traditionally long assays

The Attune NxT flow cytometer is ideal for immunophenotyping and signaling studies, cell cycle analysis, detection of rare events, stem cell analysis, cancer and apoptosis studies, microbiological assays, and more.

Fit for your lab today and in the future
The instrument’s modular design provides the flexibility to meet future capability needs with a variety of upgrade options. Purchase what you need now and easily upgrade later. The small footprint of the instrument requires minimal bench space, even allowing use in a cell culture hood.

• Flexibility—1 to 4 lasers, 14 colors, and 16 parameters
• Field upgradeable—choose to add additional lasers when you decide
• Customizable band pass filters for expanded fluorochrome choices

Tune into your data
The Attune NxT Flow Cytometer easily runs most samples including cardiomyocytes. The system is designed with a clog-resistant syringe-driven system to push the sample with a force of up to 75 psi.

• Flexibility in sample concentrations and sample types
• Elimination of time-consuming concentration steps with No-Wash, No-Lyse Filter Kit
• Analysis of dilute samples, including CSF, stem cells, mouse bone marrow, mouse blood, and thin needle aspirates
• Acquisition of a high volume of samples in minimal time

Learn what’s new with the 2019 Attune NxT cytometer and see all available configurations >
Learn more about how acoustic focusing works >
Specifications
For Use With (Equipment)Attune™ Acoustic Focusing Cytometer
FormatTube
Laser Features488 nm, 405 nm, 637 nm, 532 nm
Product LineAttune
Quantity1 each
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Unit SizeEach
Contents & Storage
• Attune Acoustic Focusing Cytometer
• Minitower Windows™ 7 64-bit computer with 23-inch flat panel monitor
• Power cord with adapters
• USB cable
• Attune NxT software license
• Quick reference card
• 1 vial of Attune Performance Tracking Beads
• 1 bottle of Attune Wash Solution
• 1 bottle of Attune 1x Shutdown Solution
• Attune Focusing Fluid (10 L)