HepExtend™ Media Kit - FAQs
HepExtend™ Media Kit - FAQs
View additional product information for HepExtend™ Media Kit - FAQs (A2933701)
2 product FAQs found
What is HepExtend Supplement?
HepExtend Supplement (50X) is an enriched supplement designed to enhance the culture lifespan (cell viability, function, and number of days in culture) of cryopreserved primary hepatocytes. We recommend using the supplement in conjunction with William's E Medium (Cat. No. A1217601) and Hepatocyte Maintenance Supplement Pack (Cat. No. CM4000), following the recommended hepatocyte culture methods. When cultured in the presence of the HepExtend Supplement, primary human hepatocyte lots will live and be functional for 10 days or longer depending on the lot of cells and other assay conditions. HepExtend Supplement does not contain growth factors, cytokines, fetal bovine serum, or pharmaceutical small molecules that are known to interfere with primary hepatocyte function.
What are the components of the HepExtend Media Kit (Cat. No.A2933701)?
The HepExtend Media Kit contains the following components needed to make the complete medium for hepatocyte culture:
- HepExtend Supplement (50X) (Cat. No.A2737501)
- Williams E Medium (500 mL) (Cat. No.A1217601)
- Primary Hepatocyte Maintenance Supplements (Cat. No.CM4000)
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.