GlycanTune™ A+ Total Feed, 100 L - FAQs
GlycanTune™ A+ Total Feed, 100 L - FAQs
View additional product information for GlycanTune™ A+ Total Feed - FAQs (A2971905, A2971904, A2971901)
5 product FAQs found
How is GlycanTune Total Feed reconstituted?
You just need to add water according to the instructions provided in the manual (https://tools.thermofisher.com/content/sfs/manuals/MAN0014673_glycantune_c_totalfeed_PI.pdf). No pH adjustment is needed
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is GlycanTune Total Feed?
It is a feed that allows you to shift the glycan profile from 90% G0F to as low as 45% G0F without significant loss of performance.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How can you support regulatory filings using GlycanTune Total Feeds?
We can provide DMF (drug master file) support or other support as needed.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the shelf life of GlycanTune Total Feeds?
The AGT dry format has a 24 month shelf-life.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
For GlycanTune Total Feeds, what do you mean by room temperature stability for 30 days post-reconstitution?
Once the product is filtered, use immediately or store at 2-8 degrees C, protected from light for up to 30 days. Our R&D team has also stored reconstituted material at 21-22 degrees C, protected from light for 30 days without any issues.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.