VetMAX™ Xeno™ Internal Positive Control DNA
Applied Biosystems™

VetMAX™ Xeno™ Internal Positive Control DNA

VetMAX™ Xeno™ Internal Positive Control (IPC) DNA serves as an internal positive control for the DNA purification process and helpsRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A29764100
A29762500
Catalog number A29764
Price (USD)
115.65
Online exclusive
128.00
Save 12.35
Each
No. of Reactions:
100
Price (USD)
115.65
Online exclusive
128.00
Save 12.35
Each
VetMAX™ Xeno™ Internal Positive Control (IPC) DNA serves as an internal positive control for the DNA purification process and helps monitor for the presence of PCR inhibitors in animal health molecular detection workflows. This is of particular importance when working with sample types that have a high level of inhibitors such as feces. Xeno IPC DNA is introduced at the nucleic acid isolation/preparation step and carried through the animal health qPCR workflow.

Features of VetMAX Xeno IPC DNA include:
• Provides confidence that qPCR test results are accurate and actionable
• Easily integrates into any workflow
• Greatly reduces the likelihood of false negatives

VetMAX Xeno IPC DNA comes in a concentration of 10,000 copies/μL and easily integrates into animal health PCR workflows, regardless of the target assay, master mix, or sample preparation reagents already in place. Coupled with a Xeno internal positive control assay, our proprietary design offers a verification layer to help ensure the PCR test results are accurate and actionable by greatly reducing the likelihood of false negatives.

Xeno IPC DNA is a component of our licensed VetMAX Gold detection kits, recommended in the American Association of Veterinary Diagnosticians guidelines, and has been successfully benchmarked against millions of genomes including those relevant to animal health. Xeno IPC RNA is also available (Cat. No. A29761).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Detection and Identification of Animal Disease
IncludesInternal Positive Control
No. of Reactions100
Product LineVetMAX, Xeno
Product TypeInternal Positive Control DNA
Quantity100 reactions
Shipping ConditionDry Ice
SpeciesNot Species-Specific
FormatKit
Unit SizeEach
Contents & Storage
  • 1 x 1250 μL Nucleic Acid Dilution Solution. Store at -5°C to -30°C.
  • 1 x 250 μL Xeno DNA Control (10,000 copies/μL). Store at -5°C to -30°C.