PureLink™ RNA Mini Kit
Invitrogen™
PureLink™ RNA Mini Kit
The PureLink RNA Mini Kit is a column-based kit used to isolate high-quality total RNA from a wide variety of sample types in 20 minutes using standard laboratory equipment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
A2983950 CartridgesRNA Mini Columns
1218302010 PrepsRNA Mini Kit
12183018A50 PrepsRNA Mini Kit
12183025250 PrepsRNA Mini Kit
1218302650 CartridgesHomogenizer
Catalog number A29839
Price (USD)
155.65
Online exclusive
164.00
Save 8.35
50 cartridges
Quantity:
50 Cartridges
Product Type:
RNA Mini Columns
Price (USD)
155.65
Online exclusive
164.00
Save 8.35
50 cartridges
The PureLink RNA Mini Kit is a column-based kit used to isolate high-quality total RNA from a wide variety of sample types in 20 minutes using standard laboratory equipment. The kit includes RNase-free lysis and wash solutions that protect RNA from RNases while liberating the RNA from DNA, proteins, and other cellular debris. The fast spin-column workflow is ideal for processing low to mid-throughput batch sizes. The advanced PureLink RNA Mini spin column design allows for maximum sample input (200 mg of tissue) and RNA recovery (up to 1000 μg). This means that users can process both small and large sample sizes with the same RNA isolation kit.
  • Process sample inputs from 5–200 mg of animal/plant tissue or from 0.5 x 106 to 108 tissue culture cells with one spin column in 20 minutes
  • Isolate more RNA than competitor mini spin columns: up to 1000 μg of total RNA from one column
  • Use isolated RNA in most applications without the need for further DNase treatment (very low minimal residual DNA carry over)
  • Flexible–optional on-column or post-isolation DNase treatment is available with PureLink DNase Set (Cat. No. 12185010) for easy removal of residual DNA
Extra-large binding capacity enables rapid RNA purification using standard laboratory equipment
The PureLink RNA Mini Kit provides rapid purification of total RNA from a wide range of cells and tissue types to yield up to 1000 μg of purified RNA from a single extraction (see Figure 1). High-quality total RNA can be obtained from mini- to midi-prep amounts of starting material with just trace amounts of residual genomic DNA contamination. The extra-large binding capacity enables one kit to handle most RNA isolation needs in a quick 20 minute protocol without the need for special sample processing instruments.
Easy, optional, on-column DNase treatment for sensitive applications
The PureLink RNA Mini Kit columns are highly efficient for isolating high quality total RNA while removing the majority of genomic DNA. In general, most applications requiring RNA from animal tissue or mammalian cell lines do not require additional DNase treatment. However, some applications such as gene expression analysis by qRT-PCR without intron-spanning primers or working with samples from organisms with very small or no introns may require more complete removal of residual contaminating DNA. The PureLink DNase Set (12185010) allows for convenient on-column digestion of DNA during the RNA isolation protocol. Treating with DNase while 'on-column' is easier and allows higher RNA recovery than treating with DNase after the RNA has been isolated. The PureLink DNase Set can also be used to remove residual DNA from RNA that has been previously purified. Both the 'on-column' and post-RNA purification workflows are options available with the PureLink DNase Set.
Simplified RNA purification
The PureLink RNA Mini Kit utilizes guanidine-isothiocyanate lysis buffer to protect the RNA during the isolation steps. Special RNase-free reagents combined with certified RNase-free silica membranes in a unique column configuration allow for a safe and easy procedure that can typically be completed in less than 20 minutes without the need of hazardous phenol/chloroform extraction, CsCl centrifugation, or LiCl or alcohol precipitation.
The PureLink RNA Mini Kit is recommended for use with the Homogenizer (Cat. No. 12183026), designed to homogenize cell or tissue lysates via centrifugation, prior to nucleic acid purification. The Homogenizer is especially effective for clarifying particulates from plant tissues.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume30 to 300 μL
Final Product TypeTotal RNA
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product TypeRNA Mini Columns
Purification Time20 min. (After sample homogenization)
Quantity50 Cartridges
ScaleMini
Shipping ConditionRoom Temperature
Starting Material AmountBacteria: ≤109 cells, Blood: ≤0.2 mL, Cells: ≤5 x 107, Plant: ≤250 mg, Tissue: ≤200 mg, Yeast: ≤5 x 108 cells, Enzymatic Reactions: ≤1.2 mL
YieldUp to 350 μg
Isolation TechnologySilica Spin Column
Sample TypeBacteria, Blood, Cells, Plant, Tissue, Yeast, Enzymatic Reactions
Unit Size50 cartridges