Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
GTF3C1 (General Transcription Factor IIIC Subunit 1) is a Protein Coding gene. Diseases associated with GTF3C1 include Nonparalytic Poliomyelitis and Bulbar Polio. Among its related pathways are Activated PKN1 stimulates transcription of AR (androgen receptor) regulated genes KLK2 and KLK3 and Gene Expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686A111; General transcription factor 3C polypeptide 1; general transcription factor IIIC, polypeptide 1 (alpha subunit); general transcription factor IIIC, polypeptide 1, alpha 220kDa; TF3C-alpha; TFIIIC 220 kDa subunit; TFIIIC box B-binding subunit; Transcription factor IIIC 220 kDa subunit; Transcription factor IIIC subunit alpha
Gene Aliases: GTF3C1; TFIIIC; TFIIIC220; TFIIICalpha
UniProt ID: (Human) Q12789
Entrez Gene ID: (Human) 2975
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support