ProFlex™ PCR System, 2 x 384-well
Applied Biosystems™

ProFlex™ PCR System, 2 x 384-well

ProFlex™ PCR Systems are the latest in our line of Applied Biosystems™ thermal cyclers, providing the reliability and performance you've come to expect from Applied Biosystems instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWarranty
A301645 years of total service coverage with 3 PMs
4484077Limited Warranty
A301633 years of total service coverage with instrument rapid exchange
A302295 years of total service coverage with instrument rapid exchange
Catalog number A30164
Price (USD)
25,710.00
Each
Warranty:
5 years of total service coverage with 3 PMs
Price (USD)
25,710.00
Each

ProFlex™ PCR Systems are the latest in our line of Applied Biosystems™ thermal cyclers, providing the reliability and performance you've come to expect from Applied Biosystems instruments. By combining an extended warranty with your instrument purchase, you'll extend the life of your investment and receive the support you need to keep your instrument running at peak performance.

Each ProFlex PCR System Extended Warranty Package includes instrument configuration and control features, as well as extended warranty benefits, that adapt to your research needs:

  • Interchange block types if your throughput needs change
  • Access the system remotely (and conveniently) through a mobile app
  • Program the instrument in seconds with a simple-to-use touchscreen interface
  • Simulate your old instrument with Thermal Simulation modes
  • Receive fast repair from factory-trained and certified service engineers
  • Minimize instrument downtime by troubleshooting with experienced Technical Support teams

For detailed instrument information and specifications, see the ProFlex PCR System (Cat. No. 4484075).

AB Repair Center (ABRC) Extended Warranty

Add additional years of warranty coverage to your existing manufacturer's limited warranty with the AB Repair Center (ABRC) extended warranty. With ABRC support, factory-trained engineers will maintain and repair your instrument at our remote service center. Just mail us your instrument and we'll maintain or repair it and ship it back to you within 3 weeks. All parts, labor, and shipping charges are included.

The ABRC extended warranty is ideal for customers in regulated environments that must maintain asset tag consistency to comply with regulated protocols.

Certain exclusions apply. Contact your Service and Support Representative for details at service.sales@thermofisher.com.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dimensions22 x 13 x 10.6 in. (56.5 x 33 x 27 cm)
Display Type8.4 in. Color TFT LCD Touchscreen
For Use With (Application)PCR
High-throughput CompatibilityHigh-throughput Compatible
Max. Ramp Rate3°C/sec. (Block), 1.6°C/sec. (Sample)
MemoryUSB and On-board
Product LineProFlex
ProgramsAuto re-start (after Power Outages), Program Overwrite Protection
QuantitySystem + 3 yr ABRC ext warranty
Reaction SpeedFast, Standard
Thermal Accuracy±0.25°C (35°C to 99.9°C)
Thermal Range0°C to 100°C
Thermal Uniformity<0.5°C (20 sec. after reaching 95°C)
User InterfaceUSB, WiFi, Cloud, Ethernet
Voltage100/240 V
Warranty5 years of total service coverage with 3 PMs
Weight45 lb. (20.4 kg)
Block Configurations2 x 384-well Isothermal Blocks
Capacity768 x 20 μL Tubes, 2 x 384-well Plate
Product TypeThermal Cycler
Unit SizeEach
Package Components
ProFlex™ 2 x 384-well PCR System
Catalog Number
4484077
Unit Size
1 instrument
Qty
1
ProFlex™ 2 x 384-well PCR System, 1 year extended warranty, ABRC Support
Catalog Number
ZG03ESCPFLEXD384W
Unit Size
3 yr ext warranty, ABRC Support
Qty
3

Frequently asked questions (FAQs)

What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf

Find additional tips, troubleshooting help, and resources within ourPCR Thermal Cyclers Support Center.