StemPro™ MSC SFM - FAQs
StemPro™ MSC SFM - FAQs
View additional product information for StemPro™ MSC SFM - FAQs (A3021701)
1 product FAQs found
Do StemPro MSC SFM Basal Medium CTS and StemPro MSC SFM Supplement CTS contain L-Glutamine?
Both CTS StemPro MSC SFM and StemPro MSC SFM Supplement (Cat. No. A1033201) do not contain L-Glutamine.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.