MicroSEQ™ ID Microbial Identification Software v3.1 Bundle - FAQs
MicroSEQ™ ID Microbial Identification Software v3.1 Bundle - FAQs
View additional product information for MicroSEQ™ ID Microbial Identification Software v3.1 Bundle - FAQs (A30371)
11 product FAQs found
You offer two MicroSEQ ID Microbial Identification Software v3.1 Bundle kits, Cat. Nos. A30371 and A38447? What is the difference between them?
MicroSEQ ID Microbial Identification Software v3.1 Bundle, Cat. No. A38447, includes all the components of Cat. No. A30371 and in addition, it includes the validated 500 16S rDNA supplemental bacterial library. Please note that Cat. No. A38447 will be replacing Cat. No. A30371.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Are additional copies of MicroSEQ libraries required for purchase when upgrading from MicroSEQ ID Microbial Identification Software v3.0 to 3.1?
Additional copies of existing libraries are not needed when upgrading to MicroSEQ ID Microbial Identification Software version 3.1.
What is the difference between MicroSEQ ID Microbial Identification Software Lite v3.1 and MicroSEQ ID Microbial Identification Software v3.1?
The Full version supports 3500 Genetic Analyzer instrument control. The Lite version supports offline analysis only.
What is the catalog number for MicroSEQ ID Microbial Identification Software v3.1 Bundle?
It is Cat. No. A30371.
Which 3500 Data Collection software version is compatible with MicroSEQ ID Microbial Identification Software v3.1?
MicroSEQ ID Microbial Identification Software v3.1 is only compatible with 3500 Data Collection software version 3.1.
There is a performance delay in analysis of the MicroSEQ ID Microbial Identification Software v3.1. How do I resolve this?
We recommend that you exit and reopen the MicroSEQ ID Software after running for a long period of time.
The display of the application audit report slows down when numerous entries exist in the application audit report of the MicroSEQ ID Microbial Identification Software v3.1. How can I resolve this delayed display issue?
Archive the audit records and delete them from the current report. They can be restored back to the report if needed.
What causes "Out of memory" issues when exporting reports in PDF format for MicroSEQ ID Microbial Identification Software v3.1?
"Out of memory" issues may occur when exporting reports in PDF format for large projects (~50 Specimen Full Gene project) analyzed with library sequence lengths exceeding 2,300 bases with 6,000 entries.
What causes "Out of memory" issues in the MicroSEQ ID Microbial Identification Software v3.1?
"Out of memory" issues may occur during analysis when library sequence lengths exceed 2,300 bases with 6,000 entries. When creating any custom library, we recommend creating ~500 bp sequence length for a 500 kit library, ~1,500 bp sequence length for a full gene kit, and ~350 bp sequence length for a fungal kit.
What genetic analysis instrument data is compatible with the MicroSEQ ID Microbial Identification Software v3.1?
Data from the following instruments are compatible with MicroSEQ ID Microbial Identification Software v3.1:
310, 3100-Avant, 3100, 3130/3130xl, 3500/3500xL, and 3730/3730xl
What are the new features for the MicroSEQ ID Microbial Identification Software v3.1?
*The software is compatible with 3500 Data Collection v3.1 software which allows 14-day on-instrument usage time for reagents.
*Improved reporting format: More information is presented on fewer pages for better printed reports.
*New "Lifetechsupport" user group has been added to assist in the service and support of the MicroSEQ Microbial ID System by a Thermo Fisher Scientific representative.
*Windows 7, 64-bit compatibility: The Lite edition of MicroSEQ ID Software is compatible with both 32- and 64-bit Windows 7 operating systems. The Full edition is compatible only with 32-bit Windows 7 operating system.