Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that belongs to a family of helicases that are involved in the ATP-dependent unwinding of nucleic acid duplexes. The encoded protein is the largest subunit of the activating signal cointegrator 1 complex that is involved in DNA repair and resistance to alkylation damage. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating signal cointegrator 1 complex subunit 3; ASC-1 complex subunit P200; ASC1p200; dJ121G13.4; dJ467N11.1; Helicase, ATP binding 1; RNA helicase family; Trip4 complex subunit p200
Gene Aliases: ASC1p200; ASCC3; HELIC1; RNAH; RQT2
UniProt ID: (Human) Q8N3C0
Entrez Gene ID: (Human) 10973
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support