3500xL Genetic Analyzer for Protein Quality Analysis - FAQs
3500xL Genetic Analyzer for Protein Quality Analysis - FAQs
View additional product information for 3500xL Genetic Analyzer for Protein Quality Analysis - FAQs (A30556)
1 product FAQs found
Is the Plate Septa, 96 well (Cat. No. 4315933) compatible with 3500/3500xL Genetic Analyzers?
Yes, it is compatible with all versions of 3500/3500xL Genetic Analyzers as well as the 3500/3500xL Dx Genetic Analyzers, specifically in RUO mode.