JetFlex™ Genomic DNA Purification Kit
Invitrogen™

JetFlex™ Genomic DNA Purification Kit

The JetFlex™ Genomic DNA Purification Kit is designed for rapid and efficient purification of genomic DNA (gDNA) from blood, bodyRead more
Have Questions?
Catalog NumberNo. of Reactions
A30701500 Preps
Catalog number A30701
Price (MXN)
-
No. of Reactions:
500 Preps

The JetFlex™ Genomic DNA Purification Kit is designed for rapid and efficient purification of genomic DNA (gDNA) from blood, body fluids (amniotic fluid, saliva, sperm, lymph), mammalian cells, tissues, plant tissues, bacteria, and yeast. Its single-tube technology allows for isolation of high quality, high molecular weight, RNA-free gDNA without the use of spin columns of a vacuum manifold.

Key benefits include:
• Large volume processing of 2 mL of whole blood or other body fluid
• Capable of processing:
– 2 x 107 mammalian cells
– 10 mg of tissue
– 500 μL bacterial culture
– 2 x 108 cells/mL yeast culture
• Simple protocol that does not require columns or vacuum manifolds

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Real-Time PCR, PCR
High-throughput CompatibilityNot High-throughput Compatible
No. of Reactions500 Preps
Product LineJetFlex
Product TypeGenomic DNA Purification Kit
Quantity500 Preps
Sample TypeBlood, Buccal Samples, Cells, Liquid Samples (e.g. Serum)
Shipping ConditionRoom Temperature
TargetGenomic DNA
FormatSpin Column
Isolation TechnologyTube Centrifugation
Unit SizeEach
Contents & Storage
• 1 x 165 mL RBC Lysis Buffer, Store at Room Temperature
• 1 x 165 mL Cell Lysis Buffer, Store at Room Temperature
• 1 x 83 mL PPT Buffer, Store at Room Temperature
• 1 x 30 mL Pellet Compactor, Store at Room Temperature
• 1 x 110 mL TE Buffer, Store (at Room Temperature
• 10 x 21 mg Proteinase K (lyophilized powder), Store 2°C to 8°C
• 5 x 1.1 mL RNase A, Store 2°C to 8°C

Frequently asked questions (FAQs)

Does the JetFlex Genomic DNA Purification Kit work with frozen blood?

Yes. However, the yield from frozen blood is significantly lower than the yield from fresh blood.