Search
| Catalog Number | Column Capacity |
|---|---|
| A30713 | 200 μL |
| A30714 | 600 μL |
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS HS50 resin, a 50-micron strong cation-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.
Features of the resin:
RoboColumn™ is a registered trademark of ATOLL GmbH.