Ion S5™ ExT Calibration Standard
Ion Torrent™
Ion S5™ ExT Calibration Standard
Have Questions?
Catalog NumberQuantity
A307381 kit
Catalog number A30738
Price (USD)
285.00
Each
Quantity:
1 kit
Price (USD)
285.00
Each
The Ion S5™ ExT Calibration Standard is a small panel consisting of known sequence content with comprehensive and uniform representation of long homopolymers up to 10-mers. The Ion S5 ExT Calibration Standard can be used in conjunction with the Ion 520™ & Ion 530™ ExT workflow for de novo research sequencing applications to allow for greater accuracy of base-calling for templates between 400 and 600 bp.
Notes:
This kit is only compatible with the Ion S5™ and Ion S5™ XL Sequencing Systems.
Use this kit with the Ion 520 & Ion 530 ExT Kit-Chef (A30670) and Torrent Suite™ Software 5.2 or higher.
Specifications
For Use With (Equipment)Ion GeneStudio S5 System, Ion S5™ XL System
Product TypeExT Calibration Standard
Quantity1 kit
Product LineIon Torrent
Software TypeTorrent Suite Software 5.2
Unit SizeEach