ProFlex™ PCR System, 2 x 96-well
Applied Biosystems™

ProFlex™ PCR System, 2 x 96-well

The ProFlex 2 x 96-well PCR system is the latest in our line of Applied Biosystems thermal cyclers. The ProFlex system combines the reliability and performance you've come to expect from Applied Biosystems instruments with the flexible configuration and control features that adapt to your research needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWarranty
A307563 years of total service coverage
4484076Limited Warranty
Catalog number A30756
Price (USD)
22,185.00
Each
Warranty:
3 years of total service coverage
Price (USD)
22,185.00
Each

The ProFlex 2 x 96-well PCR system is the latest in our line of Applied Biosystems thermal cyclers. The ProFlex system combines the reliability and performance you've come to expect from Applied Biosystems instruments with the flexible configuration and control features that adapt to your research needs.

The ProFlex 2 x 96-well PCR system allows you to:

  • Interchange the 2 x 96-well, 0.2-mL block with another block type if throughput needs change
  • Access the system remotely (and conveniently) through a mobile app
  • Program the instrument in seconds with a simple-to-use touchscreen interface
  • Simulate old instrument with Thermal Simulation modes

Interchangeable blocks

If your research requirements change in the future, or if you share the system with someone who requires a different format, simply remove the 2 x 96-well block from the top of the instrument and exchange it with another block type. This step typically takes less than 15 seconds and requires no tools or kits.

Mobile app connectivity

Have you ever set up all of your PCR reactions, only to find out that the thermal cycler is already in use? Check its availability with a mobile app before you get to the lab. Also download experimental report files, receive alerts, and more.

Simulation modes

The ProFlex PCR System is equipped with simulation modes that mimic your old thermal cycler's ramp rate. Simulation modes are available for MJ Research PTC 200, Bio-Rad™ C1000, Bio-Rad MyCycler™, Eppendorf™ Mastercycler™, Applied Biosystems 9700, Applied Biosystems 9600, and Applied Biosystems Veriti systems.

8.4-in touch screen control

The simple and intuitive touchscreen interface allows you to program with ease and limits the amount of time spent training yourself and others how to use the system.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dimensions22 x 13 x 10.6 in. (56.5 x 33 x 27 cm)
Display Type8.4 in. Color TFT LCD Touchscreen
For Use With (Application)PCR
High-throughput CompatibilityHigh-throughput Compatible
Max. Ramp Rate3°C/sec. (Block), 1.6°C/sec. (Sample)
MemoryUSB and On-board
Product LineProFlex
ProgramsAuto re-start (after Power Outages), Program Overwrite Protection
QuantitySystem + 1 yr ABRC ext warranty
Reaction SpeedFast, Standard
Thermal Accuracy±0.25°C (35°C to 99.9°C)
Thermal Range0°C to 100°C
Thermal Uniformity<0.5°C (20 sec. after reaching 95°C)
User InterfaceUSB, WiFi, Cloud, Ethernet
Voltage100/240 V
Warranty3 years of total service coverage
Weight42.5 lb. (19.3 kg)
Block Configurations2 x 96-well Isothermal Blocks
Block FormatInterchangeable
Capacity2 x 96-well, 0.2 mL Tubes
Product TypeThermal Cycler
Unit SizeEach
Package Components
ProFlex™ 2 x 96-well PCR System
Catalog Number
4484076
Unit Size
1 instrument
Qty
1
ProFlex™ 2 x 96-well PCR System, 1 year extended warranty, ABRC Support
Catalog Number
ZG03ESCPFLEXD96W
Unit Size
1 yr ext warranty, ABRC Support
Qty
1

Frequently asked questions (FAQs)

Can the ProFlex 2 x 96-well PCR System (Cat. No. 4484076, A30756) run one plate at a time?

Yes, the ProFlex 2 x 96-well PCR System can run one plate at a time.

Find additional tips, troubleshooting help, and resources within our PCR Thermal Cyclers Support Center.

What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf

Find additional tips, troubleshooting help, and resources within ourPCR Thermal Cyclers Support Center.