MetaGenID™ Software Credit
Ion Torrent™
MetaGenID™ Software Credit
MetaGenID™ cloud-based software from CosmosID enables fast, accurate, and comprehensive analyses of unassembled next-generation sequencing reads. With curated genome databasesRead more
Have Questions?
Catalog NumberQuantity
A308891 credit
Catalog number A30889
Price (USD)
302.00
Each
Quantity:
1 credit
Price (USD)
302.00
Each
MetaGenID™ cloud-based software from CosmosID enables fast, accurate, and comprehensive analyses of unassembled next-generation sequencing reads. With curated genome databases and superior sensitivity and specificity, MetaGenID Software is superior in the detection of microorganisms in metagenomic samples. CosmosID is focused on rapid identification of microorganisms for infectious disease research, public health surveillance, food safety inspections, pharmaceutical discovery, and microbiome analysis for health and wellness.
MetaGenID Software provides:
• Rapid screening of pathogens, commensals, and indicator organisms
• Comprehensive microbiome analysis
• Viral, fungal, parasite, and bacterial databases
• Simple interface with no bioinformatics experience needed
• Graphs, taxonomic trees, and other visualizations
• Batch processing
• Antibiotic resistance and virulence factors
MetaGenID Software identifies bacteria, viruses, fungi, parasites, and antibiotic resistance factors in whole-genome shotgun metagenomic samples and discriminates pathogens from their 'near neighbors' using statistical and computational methods, with no prior assumptions regarding what is present in the sample.
MetaGenID Software uses raw, unassembled reads as input and matches the sequences against the GenBook™ database, its curated reference database of known pathogens and microbes. The GenBook library constitutes hundreds of millions of marker sequences and represents both coding and non-coding sequences that are shared or uniquely identified across taxonomic levels and/or distinct nodes of a phylogenetic tree.
Purchase of MetaGenID Software credits results in the addition of these credits to your Thermo Fisher cloud account. Credits are then deducted from your account as the software is used on a credit to sample basis. See Terms and Conditions document below.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeSoftware Credit
Quantity1 credit
Sequencing TypeIon Proton™ System, Ion PGM™ System
Product LineCosmosID, MetagenID
Unit SizeEach