Ion AmpliSeq™ Direct FFPE DNA Kit, 8 preps - FAQs
Ion AmpliSeq™ Direct FFPE DNA Kit, 8 preps - FAQs
View additional product information for Ion AmpliSeq™ Direct FFPE DNA Kit - FAQs (A31133, A31136)
1 product FAQs found
With the Oncomine BRCA Research Assay, which method do you recommend for isolating gDNA?
We recommend the following DNA isolation kits:
- RecoverAll Total Nucleic Acid Isolation Kit for FFPE (Cat. No. AM1975)
- Ion Ampliseq Direct FFPE DNA Kit (Cat. Nos. A31133, A31136)
- MagMAX FFPE DNA/RNA Ultra Kit (Cat. No. A31881)
- PureLink Genomic DNA Mini Kit (Cat. Nos, K182000, K182001, K182002)