For use with QuantStudio 5 firmware version 1.5.1
Product Image
Applied Biosystems™
HID Real-Time PCR Analysis Software v1.3 Upgrade
HID Real-Time PCR Analysis Software is a tool that helps enable forensic laboratories to optimize their quantification workflow and enhance their downstream decision-making process.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A311531 License
A311545 Licenses
Catalog number A31153
Price (USD)
3,205.00
Each
Quantity:
1 License
Price (USD)
3,205.00
Each
HID Real-Time PCR Analysis Software is a tool that helps enable forensic laboratories to optimize their quantification workflow and enhance their downstream decision-making process.
The software has the following workflow features:
  • Virtual standard curve functionality
  • Customizable templates
  • Advanced data analysis capabilities including quality flags
  • STR sample normalization (dilution) and reaction setup tool
HID Real-Time PCR Analysis Software v1.3 is designed for use with the Applied Biosystems 7500 and QuantStudio 5 real-time PCR systems. It offers a set of features beyond instrument controls. HID Real-Time PCR Analysis Software v1.3 streamlines the experimental setup, supports customizable workflow templates, and enhances the data analysis and review process. It is validated for use on the Windows™ 7 platform and is may be used with all available HID Quantifiler kits.
This product includes an HID Real-Time PCR Analysis Software v1.3 Upgrade DVD and 5 user licenses to upgrade from v1.2 of the software. Documentation is not included.
Specifications
For Use With (Equipment)AmpFLSTR PCR Enhancer
Product TypeHID Real-Time PCR Analysis Software V1.3 Upgrade
Quantity1 License
Shipping ConditionRoom Temperature
Software TypeReal-Time PCR Analysis
Unit SizeEach