Oncomine™ Colon cfDNA Assay
Ion Torrent™

Oncomine™ Colon cfDNA Assay

The Oncomine Colon cfDNA Assay is part of a complete solution to detect colon (or other related gastro-intestinal) tumor-derived DNARead more
Have Questions?
Catalog NumberQuantity
A311828 Reactions
Catalog number A31182
Price (EUR)
2.100,00
Each
Quantity:
8 Reactions
Price (EUR)
2.100,00
Each

The Oncomine Colon cfDNA Assay is part of a complete solution to detect colon (or other related gastro-intestinal) tumor-derived DNA (ctDNA) in cell-free DNA (cfDNA). It provides the reagents for library construction and a single pool of multiplex PCR primers for preparation of amplicon libraries from cfDNA obtained from the plasma fraction of a single tube of whole blood. The assay enables the analysis of single nucleotide variants and short indels that are frequently mutated in colon/gastro-intestinal cancers. Fourteen genes with >240 hotspots are covered (Genes: AKT1, BRAF, CTNNB1, EGFR, ERBB2, FBXW7, GNAS, KRAS, MAP2K1, NRAS, PIK3CA, SMAD4, TP53, APC). Through the use of tag sequencing technology, a limit of detection (LOD) down to 0.1% can be achieved.

Note: Target and hotspot BED files and User Guide document for cfDNA Variant Analysis with Torrent Suite Software 5.2 can be found below under 'Documents'.

  • Optimized short amplicon design for fragmented cfDNA (∼170 bp in length)
  • Detection of both single nucleotide variants (SNVs) and hotspot indels down to 0.1% LOD
  • Very high sensitivity and specificity, enabling extremely low levels of detection with minimal false positives
  • 6 samples can be run on an Ion 530 Chip, reducing overall sequencing costs
  • Efficient workflow, from a single tube of blood to results in just 2 days

Liquid biopsies offer several advantages over conventional solid tumor biopsies:

  • Liquid biopsies are less invasive, enabling samples to be taken at multiple time points to monitor progression
  • Lower cost compared to traditional tissue biopsies
  • Faster turnaround time from sample to results
  • More tumor heterogeneity captured

The entire workflow (figure below) from isolation of cfDNA using the MagMAX Cell-Free DNA Isolation Kit to analysis of samples can be accomplished in just 2 days using the Ion S5 XL sequencing system.

Technology

ctDNA is found at extremely low concentrations in the plasma fraction of whole blood. Because of this low prevalence, a tag sequencing technology is utilized in this assay. The technology attaches a unique molecular tag to the gene-specific primers. The amplified products are then grouped into families containing the same tags. Families that contain the same mutant variant will be called with optimized Variant Caller settings for the Oncology-Liquid Biopsy application. Families that contain random errors, typically generated through the library construction/sequencing process, are identified and removed from variant calling.

Unlike other technologies with LODs of 1-5%, the Oncomine Colon cfDNA Assay has a flexible detection limit down to 0.1% or 1 mutant copy in a background of 1,000 wild-type copies. To achieve 0.1% LOD, 20 ng of input cfDNA is required. Lower amounts of cfDNA can be used (down to 1 ng), but the LOD will be higher depending on the input amount.

Simplicity, speed, and scalability of tag-sequencing technology

The Oncomine Colon cfDNA Assay enables cancer genetic studies from just 1 ng of input cfDNA for targeted library construction. The cfDNA assay uses standard PCR equipment and two simple PCR reactions, one to attach the unique molecular tags and the second to amplify the library, for high multiplex PCR-based target selection with no need for extensive capital investment. Additionally, the Oncomine Colon cfDNA Assay is compatible with FFPE samples for possible concordance studies. Total time to targeted libraries is just 3.5 hours. Scalability and flexibility are achieved using the Tag Sequencing Barcode Set 1-24 (Cat. No. A31830) or Taq Sequencing Barcode Set 25-48 (Cat. No. A31847) for multiplexing barcoded samples on Ion S5 chips.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chip CompatibilityIon 530
For Use With (Application)Oncology Research
For Use With (Equipment)Ion GeneStudio S5, Ion GeneStudio S5 Plus, Ion GeneStudio S5 Prime, Ion Chef
LibrariesAmplicon
No. of Reactions8
No. of SamplesUp to 8 Samples
Nucleic Acid TypecfDNA
Product LineOncomine
Product TypeAssay
Quantity8 Reactions
Sample TypePlasma cfDNA
Shipping ConditionDry Ice
ThroughputUp to 6 Samples per Ion 530 Chip
Workflow StepPanel and Library
Sequencing TypeNext Generation Sequencing
Unit SizeEach
Contents & Storage
Store at -30°C to -10°C.