PureLink™ Expi Endotoxin-Free Mega Plasmid Purification Kit
Invitrogen™

PureLink™ Expi Endotoxin-Free Mega Plasmid Purification Kit

The PureLink Expi Endotoxin-Free Mega Plasmid Purification Kit enables quick and simple isolation of endotoxin-free plasmid DNA using an enhancedRead more
Have Questions?
Catalog NumberQuantity
A312324 preps
Catalog number A31232
Price (USD)
448.65
Online exclusive
477.00
Save 28.35
Each
Quantity:
4 preps
Price (USD)
448.65
Online exclusive
477.00
Save 28.35
Each
The PureLink Expi Endotoxin-Free Mega Plasmid Purification Kit enables quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange membrane. In as little as 2 hours, purify up to 5 mg of high-quality plasmid DNA that is suitable for advanced transfection applications such as transfection of sensitive cells lines or genome engineering.

Advantages of the PureLink Expi Endotoxin-Free Mega Plasmid Purification Kit include:
• Simple and fast protocol—purification typically takes 2 hours
• High yield—isolate up to 5 mg of high quality plasmid DNA
• Endotoxin-free plasmid—less than 0.1 EU/μg, ideal for sensitive applications
• High purity—ultralow levels of protein, RNA, and genomic DNA contamination
• Compatibility—suitable for use with ExpiCHO and Expi293 expression systems
• Endotoxin-free components— elution buffer, water, and plasmid storage tubes that are free of endotoxin

Simple and fast protocol
Unlike other anion-exchange-based endotoxin-free plasmid purification systems, the PureLink Expi Endotoxin-Free Mega Plasmid Purification Kit utilizes centrifugation- or vacuum-assisted filtration for lysate clarification and subsequent plasmid DNA purification. There is no waiting for gravity-flow columns and the entire purification typically takes 2 hours(including the final precipitation step).

Endotoxin-free plasmid
Purify endotoxin-free (less than 0.1 EU/μg) plasmid that is ideal for sensitive applications such as transfection of primary cells, genome engineering, and research on gene therapy or plasmid vaccines in vivo.

Compatible with ExpiCHO and Expi293 expression systems
Plasmid purified using the PureLink Expi Endotoxin-Free Mega Plasmid Purification Kit is fully compatible with the Gibco ExpiCHO and Expi293 transient mammalian expression systems.

Superior quality with endotoxin-free components
Rest assured that the kit is of high quality and has additional endotoxin-free elution buffer, water, and plasmid storage tubes for added peace of mind.
For research use only. Not for use in diagnostic procedures.
Specifications
Expression SystemMammalian
For Use With (Application)Cloning, PCR
High-throughput CompatibilityManual Protocols
Prep ScaleLarge Volume Microbial Samples (> 10 mL)
Product LinePureLink
Product TypePlasmid Purification Kit
Quantity4 preps
Sample TypeBacterial Culture
Shipping ConditionRoom Temperature
TargetEndotoxin-free Plasmid
Test Time2 hr.
FormatSpin Column
Isolation TechnologyAnion Exchange Resin
Unit SizeEach
Contents & Storage

• Resuspension Buffer
• Lysis Indicator
• RNase A
• Lysis Buffer
• Precipitation Buffer
• Endotoxin Removal Buffer
• Wash Buffer
• Elution Buffer
• TE Buffer
• Expi Mega Filtration Column
• Expi Mega DNA Binding Column

Endotoxin-free components: elution buffer, water, plasmid storage tubes

Frequently asked questions (FAQs)

What makes the PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits yield endotoxin-free plasmid DNA?

The anion exchange resin used in PureLink HiPure plasmid purification kits produce plasmid DNA with inherently low endotoxin levels of 0.1-1.0 EU/µg. To add to this, PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits contain endotoxin-free components, and a proprietary Endotoxin Removal Buffer that further enhances endotoxin depletion, resulting in plasmid DNA that is consistently endotoxin-free (< 0.1 EU/µg) .

Is the Endotoxin Removal Buffer (ER) in PureLink Expi Endotoxin-Free Mega/Giga plasmid purfiication kits sold separately?

No. Endotoxin Removal Buffer (ER) in PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits is not sold separately.

What is the endotoxin level in the plasmid DNA prepared using PureLink Expi Endotoxin-Free Mega/Giga plasmid purfiication kits?

The PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits can produce endotoxin-free (< 0.1 EU/µg) advanced transfection-grade plasmid DNA.