PureLink™ Expi Endotoxin-Free Plasmid Purification Kits
Invitrogen™

PureLink™ Expi Endotoxin-Free Plasmid Purification Kits

The PureLink™ Expi Endotoxin-Free Plasmid Purification Kits enable quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantity
A31233Plasmid Purification Kit2 Preps
A31233-1RNase A1.65 mL
A31233-2Endotoxin Removal Buffer70 mL
A31233-3Lysis Indicator1 mL
A31233-4DNA Binding2 Units
A31233-5Lysate10 Units
A31233-6Tube1 Unit
A31233-7TE Buffer (TE)32 mL
A31233-8Resuspension Buffer (R3)275 mL
A31233-9Lysis Buffer (L7)280 mL
A31233-10Precipitation Buffer (N3)280 mL
A31233-11Equilibration Buffer (EQ1)450 mL
A31233-12Wash Buffer (W8)450 mL
A31233-13Elution Buffer (E4)450 mL
A31233-14Endotoxin Free Water21 mL
Catalog number A31233
Price (USD)
415.65
Online exclusive
438.00
Save 22.35
Each
Product Type:
Plasmid Purification Kit
Quantity:
2 Preps
Price (USD)
415.65
Online exclusive
438.00
Save 22.35
Each

The PureLink™ Expi Endotoxin-Free Plasmid Purification Kits enable quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange resin.

The PureLink Expi Endotoxin-Free Plasmid Purification Kits enable quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange membrane. In as little as 2 hours, purify up to 15 mg of high-quality plasmid DNA that is suitable for advanced transfection applications such as transfection of sensitive cells lines or genome engineering.

Advantages of the PureLink Expi Endotoxin-Free Plasmid Purification Kits include:

  • Simple and fast protocol—purification typically takes 2 hours
  • High yield—isolate up to 15 mg of high quality plasmid DNA
  • Endotoxin-free plasmid—less than 0.1 EU/μg, ideal for sensitive applications
  • High purity—ultralow levels of protein, RNA, and genomic DNA contamination
  • Compatibility—suitable for use with ExpiCHO and Expi293 expression systems
  • Endotoxin-free components— elution buffer, water, and plasmid storage tubes that are free of endotoxin

Simple and fast protocol

Unlike other anion-exchange-based endotoxin-free plasmid purification systems, the PureLink Expi Endotoxin-Free Plasmid Purification Kits utilize centrifugation- or vacuum-assisted filtration for lysate clarification and subsequent plasmid DNA purification. There is no waiting for gravity-flow columns and the entire purification typically takes 2 hours (including the final precipitation step).

Endotoxin-free plasmid

Purify endotoxin-free (less than 0.1 EU/μg) plasmid that is ideal for sensitive applications such as transfection of primary cells, genome engineering, and research on gene therapy or plasmid vaccines in vivo.

Compatible with ExpiCHO and Expi293 expression systems

Plasmid purified using the PureLink Expi Endotoxin-Free Plasmid Purification Kits are fully compatible with the Gibco ExpiCHO and Expi293 transient mammalian expression systems.

Superior quality with endotoxin-free components

Rest assured that the kits are of high quality and have additional endotoxin-free elution buffer, water, and plasmid storage tubes for added peace of mind.

For Research Use Only.
Specifications
Expression SystemMammalian
For Use With (Application)Cloning, PCR
High-throughput CompatibilityManual Protocols
No. of Reactions2 Preps
Prep ScaleLarge Volume Microbial Samples (> 10 mL)
Product LinePureLink
Product TypePlasmid Purification Kit
Quantity2 Preps
Sample TypeBacterial Culture
Shipping ConditionRoom Temperature
TargetEndotoxin-free Plasmid
Test Time2 hr.
FormatSpin Column
Isolation TechnologyAnion Exchange Resin
Unit SizeEach
Contents & Storage

• Resuspension Buffer
• Lysis Indicator
• RNase A
• Lysis Buffer
• Precipitation Buffer
• Endotoxin Removal Buffer
• Wash Buffer
• Elution Buffer
• TE Buffer
• Expi Giga Filtration Column
• Expi Giga DNA Binding Column

Endotoxin-free components: elution buffer, water, plasmid storage tubes

Frequently asked questions (FAQs)

What makes the PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits yield endotoxin-free plasmid DNA?

The anion exchange resin used in PureLink HiPure plasmid purification kits produce plasmid DNA with inherently low endotoxin levels of 0.1-1.0 EU/µg. To add to this, PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits contain endotoxin-free components, and a proprietary Endotoxin Removal Buffer that further enhances endotoxin depletion, resulting in plasmid DNA that is consistently endotoxin-free (< 0.1 EU/µg) .

Is the Endotoxin Removal Buffer (ER) in PureLink Expi Endotoxin-Free Mega/Giga plasmid purfiication kits sold separately?

No. Endotoxin Removal Buffer (ER) in PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits is not sold separately.

What is the endotoxin level in the plasmid DNA prepared using PureLink Expi Endotoxin-Free Mega/Giga plasmid purfiication kits?

The PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits can produce endotoxin-free (< 0.1 EU/µg) advanced transfection-grade plasmid DNA.