Pharma MagMAX™ Express-96 Deep-Well Tip Combs - FAQs
Pharma MagMAX™ Express-96 Deep-Well Tip Combs - FAQs
View additional product information for Pharma MagMAX™ Express-96 Deep-Well Tip Combs - FAQs (A31537)
1 product FAQs found
Do you offer an alternative for the discontinued Pharma MagMAX Express-96 Deep-Well Tip Combs (Cat. No. A31537)?
Yes, we recommend using KingFisher 96 tip combs for deep-well format (Cat. No. 97002534 or 97002570).