Do you offer an alternative for the discontinued Pharma MagMAX Express-96 Deep-Well Plates (Cat. No. A31540)?
Yes, we recommend using KingFisher 96 deep-well plate (Cat. No. 95040450, 95040460, or 95040455).
Search
View additional product information for Pharma MagMAX™ Express-96 Deep-Well Plates - FAQs (A31540)
1 product FAQs found