Pharma MagMAX™ Express-96 Standard Plates - FAQs
Pharma MagMAX™ Express-96 Standard Plates - FAQs
View additional product information for Pharma MagMAX™ Express-96 Standard Plates - FAQs (A31541)
1 product FAQs found
Do you offer an alternative for the discontinued Pharma MagMAX Express-96 Standard Plates (Cat. No. A31541)?
Yes, we recommend using KingFisher 96 microplate (for KingFisher Flex, Apex, and Presto) (Cat. No. 97002540).
Note: Only works with KingFisher Flex 96 Heating Block (Cat. No. 24075420). Not to be used with KingFisher Flex 96 Deep-Well Heating Block (Cat. No. 24075430).