See more signal from immunofluorescence images with Prolong® Diamond. Learn more.
BioProbes™ Journal of Cell Biology Applications—Subscribe today »
Alexa Fluor™ 594 Goat Anti-Rabbit SFX Kit, highly cross-adsorbed
Invitrogen™

Alexa Fluor™ 594 Goat Anti-Rabbit SFX Kit, highly cross-adsorbed

Alexa Fluor™ SFX Kits contain Image-iT™ FX signal enhancer (Cat. no. I36933) plus one of sixteen different Alexa Fluor™ dye-labeledRead more
Have Questions?
Catalog NumberQuantity
A316321 Kit
Catalog number A31632
Price (USD)
250.00
Each
Quantity:
1 Kit
Price (USD)
250.00
Each
Alexa Fluor™ SFX Kits contain Image-iT™ FX signal enhancer (Cat. no. I36933) plus one of sixteen different Alexa Fluor™ dye-labeled secondary antibodies. These kits provide 400 μg (0.2 mL of 2 mg/mL) of either goat anti-mouse IgG or goat anti-rabbit IgG antibody (as a standard or highly cross-adsorbed preparation) conjugated to Alexa Fluor™ 488, Alexa Fluor™ 555, Alexa Fluor™ 594, or Alexa Fluor™ 647 dye, four of our most commonly used Alexa Fluor™ dyes. The Alexa Fluor™ 594 goat anti-rabbit IgG, highly cross-adsorbed, included with this kit is also available in a 1000 μg unit size (Cat. no. A11037).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionAlexa Fluor™ 594 Highly Cross-Adsorbed Goat Anti-Rabbit SFX Kit, 1 Kit, Alexa Fluor™ 594 Conjugate, Goat Host Species, Rabbit Species Reactivity, 2 to 8°C Storage Requirements, For Antibodies and Secondary Detection, Cell Analysis
Packaging1 Kit(s)
Primary or SecondarySecondary
Product LineAlexa Fluor, Molecular Probes
ReactivityRabbit
Research DisciplineCell Analysis
Shipping ConditionRoom Temperature
Whole MoleculeWhole Antibody
ConjugateAlexa Fluor 594
Host SpeciesGoat
Quantity1 Kit
Regulatory StatusRUO, RUO
Target SpeciesRabbit
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.