Pharmaceutical Analytics QuantStudio™ 3 Real-Time PCR System, 96-well, 0.2 mL
Pharmaceutical Analytics QuantStudio™ 3 Real-Time PCR System, 96-well, 0.2 mL
Applied Biosystems™
Pharmaceutical Analytics QuantStudio™ 3 Real-Time PCR System, 96-well, 0.2 mL
The Applied Biosystems™ QuantStudio™ 3 Real-Time PCR System for Pharmaceutical Analytics is designed for users who need an affordable, easy-to-useRead more
Have Questions?
Catalog NumberQuantity
A316651 System
Catalog number A31665
Price (MXN)
-
Request A Quote
Quantity:
1 System
The Applied Biosystems™ QuantStudio™ 3 Real-Time PCR System for Pharmaceutical Analytics is designed for users who need an affordable, easy-to-use real-time PCR system that doesn't compromise performance and quality. The QuantStudio 3 pharmaceutical analytics system is used together with specialized sample preparation and quantitation kits for host cell protein, host cell DNA, and Protein A impurity testing of biological products produced in cell culture.
The simplified Design and Analysis software is ideal for both first-time and experienced users. When connected to the Thermo Fisher Cloud, the QuantStudio 3 system provides access to your data wherever and whenever you want. Using proven OptiFlex™ technology (featuring 4 coupled channels and white LED) and featuring three independent Veriflex™ temperature zones, the QuantStudio 3 system enables improved data accuracy and sensitivity for a broad range of genomic applications.
Features of the QuantStudio 3 system include:
• Simplified instrument software with interactive touchscreen for greater ease of use
• Proven OptiFlex technology (4 coupled channels with white LED) for trusted results
• Secure cloud-based software option provides increased accessibility and storage options
• User account management, locked workflow, and Pause features for greater control of experiments
• 96-well 0.2 mL format or 96-well 0.1 mL format
Thermo Fisher Cloud
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Upload data from the instrument or upload your own files from anywhere you have web access. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use.
Downloads
> QuantStudio 3/5 Design & Analysis Software | Release Notes | Example Data & Templates
> QuantStudio 3/5 Firmware | Release Notes
Easy to use
The QuantStudio 3 Real-Time PCR System is compact in form, factory calibrated and is delivered in user-friendly packaging to enable fast and simple installation for immediate use. The interactive touchscreen provides a modern usability experience and the simplified instrument software/pre-optimized protocol templates allow users of any experience level to quickly set up and start a run.
Design and analysis software options:
Option 1: QuantStudio Design and Analysis cloud software
• Supports web browser-based system configuration with PC or MAC computer
• Streamlined software for improved usability and response time
• Enables secure access of your data when and where you want through the cloud
• No software to install, no licenses and no versions to update
Option 2: QuantStudio Design and Analysis desktop software
• Supports traditional co-located computer system configuration on Windows™ 7 OS
• Streamlined software for improved usability and response time
• No licenses to purchase
Results you can trust
Improved well-to-well and instrument-to-instrument data accuracy is achieved with OptiFlex technology, which features 4 coupled excitation and emission filter channels with white LED.
Sensitive data analysis
Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
High application versatility
Simple and optimized protocols, reliable assays and reagents, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, gene regulation, and more.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Display TypeTouchscreen
Format96-well Plate
Quantity1 System
Shipping ConditionRoom Temperature
Block FormatNon-interchangeable
Unit SizeEach
Contents & Storage
• QuantStudio 3 Real Time PCR System
• 0.2 mL 96-well block
• (Computer not included)
Frequently asked questions (FAQs)
Can I downgrade the firmware on my QuantStudio 5?
You can perform this action if you have access to an Admin account on the QuantStudio instrument and if it is compatible for the downgrade (newer instruments are not compatible with v1.3.3). If your instrument serial number is below the following, then it can be downgraded:
QuantStudio 5 96-well, 0.1 mL block - SN 272514140
QuantStudio 5 96-well, 0.2 mL block - SN 2725210507
QuantStudio 5 384 well block - SN 272533285
Please also check if the instrument is being used with any validated protocols that may require a particular firmware version to be compliant. If you need the firmware itself, you can download it here.
The instructions to downgrade the firmware are on p.4 of the Release Notes: QuantStudioTM 3 & 5 Touchscreen Instrument Operating Software Version 1.3.3. It's the same process as an upgrade.
What is the admin pin for QuantStudio 3 and 5 Real-Time PCR Systems?
If the admin PIN has not been changed, 1234 should work. If it does not work, you can also try 0000.
However, if the admin PIN has been changed and none of the above PINs work, follow the instructions in the QuantStudio 3 and 5 Real-Time PCR Systems manual, page 100.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.