Pharmaceutical Analytics QuantStudio™ 5 Real-Time PCR System, 384-well, laptop - FAQs
Pharmaceutical Analytics QuantStudio™ 5 Real-Time PCR System, 384-well, laptop - FAQs
View additional product information for Pharmaceutical Analytics QuantStudio™ 5 Real-Time PCR System, 384-well, laptop - FAQs (A31677)
2 product FAQs found
I am unable to download the current software update for the QuantStudio 5 Real-Time PCR System from the Thermo Fisher website. Why?
We recommend trying different browsers (Google Chrome, Firefox, and Microsoft Edge) and/or removing cookies from your browser. If this does not resolve the issue, please contact Technical Support at techsupport@thermofisher.com.