MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plates with Barcode, blue
The Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plate is optimally designed for use with the Applied Biosystems™ Automated ThermalRead more
Have Questions?
Catalog Number
Quantity
A31727
50 plates
Catalog number A31727
Price (USD)
350.65
Online exclusive
377.00
Save 26.35
Each
Quantity:
50 plates
Price (USD)
350.65
Online exclusive
377.00
Save 26.35
Each
The Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Full-Skirted Plate is optimally designed for use with the Applied Biosystems™ Automated Thermal Cycler (ATC). These full-skirted reaction plates are manufactured using a durable polycarbonate frame, making them ideal for easy handling with robotic arms and other automated system features. The thin-wall construction of the plate allows for efficient heat transfer and optimal fit in thermal cycler blocks. The black alpha- numeric labeling provides easy-to-read well identification during sample preparation. The MicroAmp EnduraPlate Optical 96-Well Full-Skirted Plate is the only 96-well plate recommended for use with the Automated Thermal Cycler (ATC) in the automated PCR workflow process.
Key features include: • Designed exclusively for use with Automated Thermal Cycler (ATC) (Cat No. A31486) • Full-skirted polycarbonate frame for easy robotic handling and plate stacking/loading • 3-way barcodes on alpha, opposite alpha, and opposite numeric sides to improve plate reading efficiency • Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and automated work processes • Certified free of DNA, RNase, and PCR inhibitors • Available in multiple colors for easy organization of laboratory assays • Excellent mechanical stability and flatness for your complete automated workflow needs
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer