Spectral Calibration Plate 3 (TAMRA™, NED™, CY™5), 96-well
Applied Biosystems™

Spectral Calibration Plate 3 (TAMRA™, NED™, CY™5), 96-well

The Spectral Calibration Plate 3 (TAMRA™, NED™, CY™5), 96-well is a specialized calibration tool to establish pure dye spectra and multicomponent values for three fluorescent dyes: TAMRA, NED, CY5.
Have Questions?
KatalognummerMenge
A319091 Plate
Katalognummer A31909
Preis (EUR)
206,04
Exklusiv online
404,00
Ersparnis 197,96 (49%)
Each
Menge:
1 Plate
Preis (EUR)
206,04
Exklusiv online
404,00
Ersparnis 197,96 (49%)
Each

The Spectral Calibration Plate 3 (TAMRA™, NED™, CY™5), 96-well is a specialized calibration tool to establish pure dye spectra and multicomponent values for three fluorescent dyes: TAMRA, NED, CY5. This multi-dyeplate offers significant operational efficiency by enabling simultaneous calibrations of multiple dyes within a single 96-well format, substantially reducing the effort and time traditionally required for individual dye calibration procedures.

Features:

  • Compatible with QuantStudio 5 Dx and QuantStudio 7 Pro Dx 96-well real-time PCR systems
  • Multiple Dye Compatibility: Supports simultaneous detection using multiple fluorescent dyes, enabling multiplex assay capabilities for enhanced analytical throughput

Simultaneous Multi-Dye Calibration Capability

This qPCR-based calibration system advances fluorescent dye calibration by enabling simultaneous calibration of multiple dyes in a single plate, reducing the time and effort required compared to traditional sequential methods.

Single-Plate Format Optimizes Resource Utilization

The single-plate format improves efficiency by enabling comprehensive multi-dye calibrations instead of multiple plates, while also reducing inter-plate variability to enhance calibration consistency.

Usage Guidelines and Technical Requirements

Proper use of this spectral calibration system requires following the procedures outlined in the instrument user guide to enable optimal performance, validated calibration, and compliance with quality standards. Users should consult the complete instrument documentation to achieve optimum performance and maintain compliance with established quality standards for qPCR spectral calibration procedures.

For In Vitro Diagnostic Use.
Specifications
Kalibrierter FarbstoffTAMRA, NED, CY5
KalibrierungsartSpectral Calibration Plate
Zur Verwendung mit (Anwendung)Real-Time PCR (qPCR)
Zur Verwendung mit (Geräte)QuantStudio 5 Dx and QuantStudio 7 Pro Dx
Plattenkompatibilität96-well
Menge1 Plate
VersandbedingungDry Ice
ProdukttypDye Spectral Calibration Plate
Unit SizeEach
Inhalt und Lagerung
Store at -20°C ±5°C.