TaqMan™ Vaginal Microbiota Amplification Control, 20 μL - FAQs
TaqMan™ Vaginal Microbiota Amplification Control, 20 μL - FAQs
View additional product information for TaqMan™ Vaginal Microbiota Amplification Control - FAQs (A32040)
2 product FAQs found
Do you have any positive controls for the vaginal microbiota assays?
The TaqMan Vaginal Microbiota Amplification Control (Cat. No. A32040) is a linearized multi-target plasmid with target sequences for each available vaginal microbiota profiling assay.
Which sample prep kit do you recommend when running vaginal microbiota profiling experiments?
We recommend the MagMAX DNA Multi-Sample Ultra Kit (Cat. No. A25597) for isolating DNA for vaginal microbiota profiling experiments.