Product Image
Pentosidine (CAY-10010254-10mg)
Have Questions?
Catalog number A32246
Price (USD)
-
Pentosidine
Specifications
Product Size10mg
Third-Party BrandsCayman Chemical Company 제조, Life Technologies Korea LLC 판매 및 배송
Unit Size10mg
Frequently asked questions (FAQs)
Can I store the MicroSEQ ID Ultra Sequencing Clean-up 8-Strips Kit (Cat. No. A33246) at room temperature (20-24 degrees C)?
Yes. The kit contents are stable when stored in their original packaging at room temperature for up to 4 weeks. However, the recommended storage condition is at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.