Orbitor™ RS2 Microplate Mover for QuantStudio™ Systems, Random Access - FAQs
Orbitor™ RS2 Microplate Mover for QuantStudio™ Systems, Random Access - FAQs
View additional product information for Orbitor™ RS2 Microplate Mover for QuantStudio™ Systems, Random Access - FAQs (A32269)
1 product FAQs found
Is the ViiA7 Real-Time PCR System compatible with the 7900HT Fast System Twister robot?
No, the ViiA7 Real-Time PCR System is not compatible with the 7900HT Fast System Twister robot. For automation, the ViiA7 Real-Time PCR System can be connected to one of the following robots:
• Orbitor RS2 Microplate Mover for QuantStudio Systems, Sequential Access (cat. No. A32268)
• Orbitor RS2 Microplate Mover for QuantStudio Systems, Random Access (cat. No. A32269)
• Twister Automation Robot for QuantStudio Systems (Cat. No. 4471066)
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.