Do the master mixes included in the HIV-1 Genotyping Kit Amplification Module (Cat. No. A32317) contain the necessary primers?
Yes, we provide the primers in the master mixes. They are not in separate tubes.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.