KingFisher™ Flex Purification System with 96 Deep-Well Head - FAQs
KingFisher™ Flex Purification System with 96 Deep-Well Head - FAQs
View additional product information for KingFisher™ Flex Purification System with 96 Deep-Well Head - FAQs (A32681)
2 product FAQs found
Can I re-start a script that has stopped mid-way on the KingFisher Flex/KingFisher Duo Prime instrument?
The only way is to do this is by rewriting the script from where you want it to start, or start the run all over again.
I have a BindiT protocol installed on the KingFisher Flex/DuoPrime instrument and would like to edit it. How do I do it?
There is no way to edit a BindiT protocol that has been installed on the KingFisher Flex/KingFisher DuoPrime instrument. The only way to edit the protocol is by saving the original .bdz file to a PC and then editing it in BindiT software.