When using the MagMAX CORE Nucleic Acid Purification Kit (Cat. No. A32700), I am getting good DNA / RNA concentrations and also acceptable 260/280 ratios, but, however, very low 260/230 ratios (<1), how could I improve this value to be closer to 1.8?
Performing an additional alcohol wash step will help improve this ratio.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.