MagMAX™ CORE Nucleic Acid Purification Kit, 500 rxns - FAQs
MagMAX™ CORE Nucleic Acid Purification Kit, 500 rxns - FAQs
View additional product information for MagMAX™ CORE Nucleic Acid Purification Kit - FAQs (A32700, A32702)
3 product FAQs found
With the MagMAX CORE Nucleic Acid Purification Kit, do I need to add alcohol to any of the wash buffers?
No. All buffers arrive pre-mixed with alcohol.
Where can I find the .bdz files for use with the MagMAX CORE Nucleic Acid Purification Kit?
You can find the .bdz files located within the Documents section of the MagMAX CORE product page (https://www.thermofisher.com/order/catalog/product/A32700) under "Product Literature".
What is different about the MagMAX CORE Nucleic Acid Purification Kit compared to the other MagMAX kits?
The MagMAX CORE Nucleic Acid Purification Kit offers pre-mixed, room-temperature reagents, which greatly improve product handling and storage. Specifically, the CORE kit decreases processing times by having pre-mixed washes that do not require the addition of alcohol, a single protocol adapted on multiple KingFisher magnetic particle processors, and a complete protocol that addresses 14 unique sample types.